missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iso Furosemide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-4-[(furan-2-ylmethyl)-amino]-5-sulfamoylbenzoic Acid |
| CAS | 4818-59-1 |
| Molecular Formula | C12 H11 Cl N2 O5 S |
| Quantity | 100 mg |
| Synonym | 2-Chloro-4-[(furan-2-ylmethyl)-amino]-5-sulphamoylbenzoic Acid, Furosemide Imp. A (EP) |
| InChI Formula | InChI=1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)11(21(14,18)19)4-8(9)12(16)17/h1-5,15H,6H2,(H,16,17)(H2,14,18,19) |
| SMILES | NS(=O)(=O)c1cc(C(=O)O)c(Cl)cc1NCc2occc2 |
| IUPAC Name | 2-chloro-4-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid |
| Molecular Weight (g/mol) | 330.74 |
| Formula Weight | 330.0077 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?