missing translation for 'onlineSavingsMsg'
Learn More

Iso-FK-506 (iso-Tacrolimus), TRC

Product Code. 30012665
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
10 mg
Unit Size:
0.5mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012665 10 mg 10mg
30010031 5 mg 5mg
30006148 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012665 Supplier Toronto Research Chemicals Supplier No. TRCI81625010MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134590-88-8
Molecular Formula C44H69NO12
Molecular Weight (g/mol) 804.02
Synonym [3S-[3R*[1R*,2R*,4(1S*,3S*,4S*)],6S*,10R*,12R*,13R*,14R*,16S*,17S*,24aR*]]-6,9, 10,11,12,13,14,15,16,17,22,23,24,24a-tetradecahydro-17-hydroxy-3-[2-hydroxy-4-(4-hydroxy-3-methoxycyclohexyl)-1,3-dimethyl-3-butenyl]-12,14-dimethoxy-8,10,16-trimethyl-6-(2-propenyl)-13,17-epoxy-3H-pyrido[2,1-c][1,4]oxaazacycloheneicosine-1,5,18,19(4H,21H)-tetrone, Tacrolimus Lactone Isomer
SMILES O[C@H]1[C@H](OC)C[C@H](/C=C([C@@H](O)[C@H](C)[C@@H](O2)CC([C@H](CC=C)/C=C(C)/C[C@H](C)C[C@H](OC)[C@](O3)([H])[C@@H](OC)C[C@@H](C)[C@]3(O)C(C(N4CCCC[C@H]4C2=O)=O)=O)=O)\C)CC1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C44H69NO12/c1-10-13-31-19-25(2)18-26(3)20-37(54-8)40-38(55-9)22-28(5)44(52,57-40)41(49)42(50)45-17-12-11-14-32(45)43(51)56-35(24-34(31)47)29(6)39(48)27(4)21-30-15-16-33(46)36(23-30)53-7/h10,19,21,26,28-33,35-40,46,48,52H,1,11-18,20,22-24H2,2-9H3/b25-19+,27-21+/t26-,28+,29+,30-,31+,32-,33+,35-,36+,37-,38-,39+,40+,44+/m0/s1
Percent Purity >95%
Chemical Name or Material Iso-FK-506 (iso-Tacrolimus)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.