missing translation for 'onlineSavingsMsg'
Learn More

Iso Cytosine, TRC

Product Code. 30389016
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389016 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389016 Supplier Toronto Research Chemicals Supplier No. TRCI8125005G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 108-53-2
Molecular Formula C4 H5 N3 O
Quantity 5 g
Synonym Isocytosine
InChI Formula InChI=1S/C4H5N3O/c5-4-6-2-1-3(8)7-4/h1-2H,(H3,5,6,7,8)
SMILES NC1=NC=CC(=O)N1
IUPAC Name 2-amino-1H-pyrimidin-6-one
Molecular Weight (g/mol) 111.1
Formula Weight 111.0433
Chemical Name or Material Isocytosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.