missing translation for 'onlineSavingsMsg'
Learn More

Iso Cyclosporin A, TRC

Product Code. 18090230
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18090230 5 mg 5mg
18080070 25 mg 25mg
18020450 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18090230 Supplier Toronto Research Chemicals Supplier No. TRCI8115005MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59865-16-6
Molecular Formula C62 H111 N11 O12
Molecular Weight (g/mol) 1202.61
Synonym Isociclosporin A, (2S,3R,4R,6E)-3-Hydroxy-4-methyl-2-(methylamino)-6-octenoyl-(2S)-2-aminobutanoyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl-L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valine (11→1)-lactone, Isocyclosporin A, 1-Oxa-4,7,10,13,16,19,22,25,28,31-decaazacyclotetratriacontane, cyclic peptide deriv., [2S-(2R*,3S*,4S*,6E)]-N-[3-Hydroxy-4-methyl-2-(methylamino)-1-oxo-6-octenyl]-L-alpha-aminobutyryl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl-L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valine sigma1-lactone
IUPAC Name (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S,34R)-30-ethyl-34-[(E,2R)-hex-4-en-2-yl]-4,7,10,12,15,19,25,28-octamethyl-33-(methylamino)-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1-oxa-4,7,10,13,16,19,22,25,28,31-decazacyclotetratriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone
SMILES CC[C@@H]1NC(=O)[C@@H](NC)[C@H](OC(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C)[C@H](C)C\C=C\C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C62H111N11O12/c1-25-27-28-40(15)52-50(63-18)56(78)66-43(26-2)58(80)68(19)33-48(74)69(20)44(29-34(3)4)55(77)67-49(38(11)12)61(83)70(21)45(30-35(5)6)54(76)64-41(16)53(75)65-42(17)57(79)71(22)46(31-36(7)8)59(81)72(23)47(32-37(9)10)60(82)73(24)51(39(13)14)62(84)85-52/h25,27,34-47,49-52,63H,26,28-33H2,1-24H3,(H,64,76)(H,65,75)(H,66,78)(H,67,77)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1
Formula Weight 1201.8414
Percent Purity >95%
Chemical Name or Material Isocyclosporin A
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.