missing translation for 'onlineSavingsMsg'
Learn More

Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk, Fisher Chemical™

Product Code. 16283951 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
500 mL
Packaging:
Amber Glass Bottle
Plastic Coated Glass Bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16283951 1 L Amber Glass Bottle 1 L
10409360 500 mL Amber Glass Bottle 500mL
10152040 2.5 L Plastic Coated Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16283951 Supplier Fisher Chemical Supplier No. A/7000/15
  • ONLINE EXCLUSIVE PRICE
    £80.10
    £78.70 / 1 L
    Save £1.40
    1% Off
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Iso-Amyl Alcohol, Extra Pure, SLR, for Determination Of Fat In Milk

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Weight (g/mol) 88.15
MDL Number 2934
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Melting Point -117°C
pH 6.5
Boiling Point 130°C
Packaging Amber Glass Bottle
Molecular Formula C5H12O
Quantity 1 L
Vapor Density 3.04
Formula Weight 88.15g/mol
Vapor Pressure 4 hPa at 20°C
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.