missing translation for 'onlineSavingsMsg'
Learn More

ISO 9377 Quality Control Standard 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30313416
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313416 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313416 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000973AC
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mineral Oil
CAS 8012-95-1
Analyte or Component Names Mineral Oil
Shipping Condition Ice pack (-20°C)
Concentration 500 μg/mL
Certifications/Compliance ISO 17034
Synonym Paraffin oil, Acme S 105, Actrel 3349L, Actrel 3360L, Actrel 4493L, Additol VXW 4932, Agrirob;Ampol C15 CPD, Ampol C23 DC-Tron NR, Arcoprime 200, Atpolan;Atpolan 80EC, Avilub RS, BP Energol GR-XP 220, BP Energol WM 2, Banole, Banole HV, Barrel ProcessOil P 10000, Barrel Process Oil P 2200, Cactus Normal Paraffin YHNP, Caloria HT 43, Calpar 70, Cantenex SM 925, Carnation, Certrex 60, Chevron 100R, Citation 70NY, Citgo 340 Neutral, Citrole, Clenvex AS 58;Coker gatch oil, Conosol C 200, Cosmo Neutral 500, Crop oil, Crystal Plus 70T, Cypar 7, Dalian SN 70, Diana Fresia P 430, Dicera 11833, Drakeol, Duoprime;Edelex SM 925, Elevast, Enerpar, Escaid 90, Essobayol 90, Essotherm 500;Exchange, Exssol D 80, Extensoil 51-24T, Exxal, Exxsol, Exxysn, Finavestan A;Fixaoil, Flavex 913, Flexon, Fluid paraffins;Fractol A, Freezene, Fukkol Process Oil P 100, Hiclean CF 50, Hicol, Hydrorafinat H 5, Hydroseal G, Hydrowax, Hyper White 350, Hyprene P 150BS, Iberpesa T 601, Inland 19, Ipsol H, Irgawax;Isocer NL, JMS Stylet-Oil, JOMO Process, Kemesol, Kermac 500T, Kixx LP, Klearol, Klearol 50, Kolex 320, Komorex, Kornoil Concentrate, Kyowazol C 1200, Kyowazol C 900, Lacolene, Lacquer Diluent 6, Lamacit APM 520, Liquid Paraffin 60S, Lubinol, Lucant 360;MagieSol 44, Magnaflux, Manol, Marcol, Markol 82, Michang P 3, Miclube 1460, Mineral Seal Oil, Mineral oil, paraffinic, Mobilcer, Mobiltherm 603;Moresco P 40, Nisseki Hi-White 70, Nopco 1407, Normal Paraffin TS 6, Normal paraffin, NovaSpec 2050, Nujol, O-WO2, Octafoam, Octopus P Plus 3029, Oleo FC, Ondina 15;Organic JMS Stylet-Oil, Osco Baby Oil, Pale Oil 60;Parabase, Paracos KF 350, Paraffin, Paraflex, Paralux, Paramil MES 15, Parax 40K3, Parex II, Parol 70, Parprol 22;Parprol 32, Peneteck, PennzUltra, Peptoil, Perleam 18, Petrolatum oil;Petrolatum, liq., Pharma 240B/HV6TI, Phazol 35, Pioneer 2002, Pionier 2024, Plastol 355, Plastol 537, Plaxene 185, Primol, Process oils;
Solvent or Matrix Acetone
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.