missing translation for 'onlineSavingsMsg'
Learn More

ISO 9377 Extraction Solvent Stock Solution, Dr. Ehrenstorfer

Product Code. 30325138
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325138 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325138 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000974HE

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣Tetracontane
CAS 124-18-5, 4181-95-7
Analyte or Component Names n-Decane∣Tetracontane
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Hexane
Quantity 10 mL
Concentration or Composition (by Analyte or Components) n-Decane 14.6 μg/mL∣Tetracontane 20 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.