missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Ismatec™ Silicone Oil Spray

Product Code. 10363462
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10363462

Brand: Ismatec™ SC0179A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Silicone Oil Spray
For Use With (Application) Lubrication
Quantity 1 Ea.
compliance-icons
Product Identifier
  • UNISILKON M 2000 Spray
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 1
Hazard Statement
  • H222-Extremely flammable aerosol.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P251-Do not pierce or burn, even after use.
  • P211-Do not spray on an open flame or other ignition source.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.