missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21540-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a type 1 thrombospondin domain, which is present in thrombospondin, a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
| ISM2 | |
| Polyclonal | |
| Unconjugated | |
| ISM2 | |
| ISM2, Isthmin 2, isthmin 2 homolog (zebrafish), TAIL1, THSD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 145501, 217738 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6H9L7 | |
| Ism2 | |
| ISM2 Fusion Protein Ag16187 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction