missing translation for 'onlineSavingsMsg'
Learn More

ISLR-2 Antibody [Biotin], Novus Biologicals™

Product Code. 30734247 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734247 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734247 Supplier Novus Biologicals Supplier No. AF4650B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ISLR-2 Polyclonal antibody specifically detects ISLR-2 in Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISLR-2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias immunoglobulin superfamily containing leucine-rich repeat 2, KIAA1465, leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein, LINX, UNQ1885/PRO4329
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ISLR-2, Cys20-Leu590, Accession # Q5RKR3
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57611
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.