missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567377
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MVDIIFHYPFL GAMGDHSKKK PGTAMCVGCG SQIH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
ISL2 is a transcriptional factor that defines subclasses of motoneurons that segregate into columns in the spinal cord and select distinct axon pathways.Specifications
ISL2 | |
Polyclonal | |
Unconjugated | |
ISL2 | |
3110001N10Rik; Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL LIM homeobox 2; ISL2; Isl-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; islet-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96A47 | |
ISL2 | |
Recombinant Human ISL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only