missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ISL2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA535175

Product Code. 15387584

  • £440.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

ISL LIM Homeobox 2 (ISL2) is a gene located on chromosome 15q23 that encodes a transcription factor belonging to the LIM-homeodomain family. These transcription factors play critical roles in the regulation of gene expression during embryonic development, particularly in the differentiation and maintenance of neuronal and pancreatic cell lineages. ISL2 is specifically involved in the development of the nervous system, where it contributes to motor neuron differentiation and connectivity, as well as in the development of certain retinal structures. The protein functions by binding to specific DNA sequences, thereby influencing the transcription of target genes essential for cellular differentiation and identity. Although less studied than its paralog ISL1, ISL2's expression and roles in developmental pathways are crucial for proper organogenesis and tissue specification. Dysregulation of ISL2 has potential implications in developmental disorders and diseases involving neural and pancreatic tissues. Ongoing research aims to further elucidate its functions and potential as a therapeutic target in developmental biology and disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISL2
Polyclonal
Unconjugated
ISL2
3110001N10Rik; Insulin gene enhancer protein ISL-2; insulin related protein 2 (islet 2); ISL LIM homeobox 2; ISL2; Isl-2; ISL2 transcription factor, LIM/homeodomain, (islet-2); islet 2; islet-2
Rabbit
Antigen affinity chromatography, Protein A
RUO
64843
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.4 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q96A47
ISL2
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human ISL2.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.