missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ishikawa's Reagent, 90+%

Product Code. 11399365
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399365 25 g 25g
11309375 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399365 Supplier Thermo Scientific Chemicals Supplier No. L16738.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 309-88-6 | C7H11F6N | 223.162 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 309-88-6
Molecular Formula C7H11F6N
Molecular Weight (g/mol) 223.162
MDL Number MFCD00054683
InChI Key BNTFCVMJHBNJAR-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
PubChem CID 136149
IUPAC Name N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
SMILES CCN(CC)C(C(C(F)(F)F)F)(F)F

Specifications

Density 1.23
Boiling Point 56°C to 57°C (58 mmHg)
Flash Point 14°C (57°F)
Linear Formula CF3CFHCF2N(CH2CH3)2
Refractive Index 1.35
Quantity 25 g
UN Number UN2924
Formula Weight 223.16
Percent Purity ≥90%
Chemical Name or Material Ishikawa's Reagent

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.