missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISG15/UCRP Rabbit anti-Human, Clone: 239, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £458.00
Specifications
| Antigen | ISG15/UCRP |
|---|---|
| Clone | 239 |
| Dilution | Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227166
|
Novus Biologicals
NBP2-90146-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18214488
|
Novus Biologicals
NBP2-90146-100UL |
100 μg |
£458.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ISG15/UCRP | |
| Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17 | |
| Recombinant Human ISG15/UCRP protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 239 | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway | |
| 0.2 um filtered solution in PBS | |
| 9636 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title