missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ISG15/UCRP Monoclonal antibody specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Purification Method | Protein A purified |
| Research Discipline | Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?