Learn More
ISG15/UCRP Antibody (239), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90146MFV500
Product Code. 18539581
  • £510.00 / 0.10mL
Description
ISG15/UCRP Monoclonal antibody specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50mM Sodium Borate
Purified