missing translation for 'onlineSavingsMsg'
Learn More

ISG15/UCRP Antibody (239), DyLight 680, Novus Biologicals™

Product Code. 18327050 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327050 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327050 Supplier Novus Biologicals Supplier No. NBP290146FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISG15/UCRP
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 239
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ISG15/UCRP (AAH09507.1; Met1-Ser165).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9636
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.