Learn More
ISG15 Mouse, anti-Human, Clone: 3G9A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68408-1-IG-150UL
This item is not returnable.
View return policy
Description
The ISG15 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ISG15. This antibody recognizes human antigen. The ISG15 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ISG15 | |
| 3G9A2 | |
| G1P2, hUCRP, IFI15, IP17, ISG15, ISG15 ubiquitin like modifier, Ubiquitin like protein ISG15, UCRP | |
| 150 μL | |
| 9636 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |