missing translation for 'onlineSavingsMsg'
Learn More

ISG15 Activating Enzyme/UBE1L Antibody, Novus Biologicals™

Product Code. 18392928 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392928 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392928 Supplier Novus Biologicals Supplier No. H00007318B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ISG15 Activating Enzyme/UBE1L Polyclonal antibody specifically detects ISG15 Activating Enzyme/UBE1L in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ISG15 Activating Enzyme/UBE1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH06378
Gene Alias D8UBA1, ubiquitin-activating enzyme E1 homolog B, UBA1B, UBA7, ubiquitin-activating enzyme E1, UBE1L, UBE2MGC12713, Ubiquitin-activating enzyme 7, Ubiquitin-activating enzyme E1 homolog, ubiquitin-activating enzyme E1-like, ubiquitin-activating enzyme E1-related protein, ubiquitin-activating enzyme-2, ubiquitin-like modifier activating enzyme 7, ubiquitin-like modifier-activating enzyme 7
Host Species Mouse
Immunogen UBE1L (AAH06378, 1 a.a. - 1012 a.a.) full-length human protein. MDALDASKLLDEELYSRQLYVLGSPAMQRIQGARVLVSGLQGLGAEVAKNLVLMGVGSLTLHDPHPTCWSDLAAQFLLSEQDLERSRAEASQELLAQLNRAVQVVVHTGDITEDLLLDFQVVVLTAAKLEEQLKVGTLCHKHGVCFLAADTRGLVGQLFCDFGEDFTVQDPTEAEPLTAAIQHISQGSPGILTLRKGANTHYFRDGDLVTFSGIEGMVELNDCDPRSIHVREDGSLEIGDTTTFSRYLRGGAITEVKRPKTVRHKSLDTALLQPHVVAQSSQEVHHAHCLHQAFCALHKFQHLHGRPPQPWDPVDAETVVGLARDLEPLKRTEEEPLEEPLDEALVRTVALSSAGVLSPMVAMLGAVAAQEVLKAISRKFMPLDQWLYFDALDCLPEDGELLPSPEDCALRGSRYDGQIAVFGAGFQEKLRRQHYLLVGAGAIGCELLKVFALVGLGAGNSGGLTVVDMDHIERSNLSRQFLFRSQDVGRPKAEVAAAAARGLNPDLQVIPLTYPLDPTTEHIYGDNFFSRVDGVAAALDSFQARRYVAARCTHYLKPLLEAGTSGTWGSATVFMPHVTEAYRAPASAAASEDAPYPVCTVRYFPSTAEHTLQWARHEFEELFRLSAETINHHQQAHTSLADMDEPQTLTLLKPVLGVLRVRPQNWQDCVAWALGHWKLCFHYGIKQLLRHFPPNKVLEDGTPFWSGPKQCPQPLEFDTNQDTHLLYVLAAANLYAQMHGLPGSQDWTALRELLKLLPQPDPQQMAPIFASNLELASASAEFGPEQQKELNKALEVWSVGPPLKPLMFEKDDDSNFHVDFVVAAASLRCQNYGIPPVNRAQSKRIVGQIIPAIATTTAAVAGLLGLELYKVVSGPRPRSAFRHSYLHLAENYLIRYMPFAPAIQTFHHLKWTSWDRLKVPAGQPERTLESLLAHLQEQHGLRVRILLHGSALLYAAGWSPEKQAQHLPLRVTELVQQLTGQAPAPGQRVLVLELSCEGDDEDTAFPPLHYEL
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7318
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.