missing translation for 'onlineSavingsMsg'
Learn More

Isethionic acid sodium salt, 98%

Product Code. 11458990
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458990 1000 g 1000g
11468990 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458990 Supplier Thermo Scientific Alfa Aesar Supplier No. A12054.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1562-00-1 | C2H5NaO4S | 148.108 g/mol

Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps. It makes a dense lather in addition to the lather made by the soap. It is mild on the skin, and non-drying. It works equally well in soft or hard water. It is also an anti-static agent in shampoos.

Solubility
Soluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1562-00-1
Molecular Formula C2H5NaO4S
Molecular Weight (g/mol) 148.108
MDL Number MFCD00007534
InChI Key LADXKQRVAFSPTR-UHFFFAOYSA-M
Synonym sodium 2-hydroxyethanesulfonate, sodium isethionate, isethionic acid sodium salt, 2-hydroxyethanesulfonic acid sodium salt, sodium hydroxyethylsulfonate, ethanesulfonic acid, 2-hydroxy-, monosodium salt, sodium 2-hydroxyethylsulfonate, sodium 2-hydroxyethanesulphonate, sodium beta-hydroxyethanesulfonate, sodium 1-hydroxy-2-ethanesulfonate
PubChem CID 517063
IUPAC Name sodium;2-hydroxyethanesulfonate
SMILES C(CS(=O)(=O)[O-])O.[Na+]

Specifications

Melting Point 191°C to 194°C
Quantity 1000 g
Beilstein 3633992
Solubility Information Soluble in water.
Formula Weight 148.11
Percent Purity 98%
Chemical Name or Material Isethionic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.