missing translation for 'onlineSavingsMsg'
Learn More

Isepamicin Sulfate, TRC

Product Code. 30003909
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003909 2.5 g 2.5g
30005278 500 mg 500mg
30011242 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003909 Supplier Toronto Research Chemicals Supplier No. TRCI9270132.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67814-76-0
Molecular Formula C22H43N5O12 • H2O4S
Molecular Weight (g/mol) 569.609808
Synonym D-Streptamine, O-6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)]-N1-(3-amino-2-hydroxy-1-oxopropyl)-2-deoxy-, (S)-, sulfate (salt), D-Streptamine, O-6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-L-arabinopyranosyl-(1→6)]-N1-[(2S)-3-amino-2-hydroxy-1-oxopropyl]-2-deoxy-, sulfate (salt) (9CI), Exacin, Isepacin
SMILES CN[C@@H]1[C@@H](O)[C@H](OC[C@]1(C)O)O[C@@H]2[C@@H](O)[C@H](O[C@H]3O[C@H](CN)[C@@H](O)[C@H](O)[C@H]3O)[C@@H](N)C[C@H]2NC(=O)[C@@H](O)CN.O[S](O)(=O)=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C22H43N5O12.H2O4S/c1-22(35)6-36-20(15(33)18(22)26-2)39-17-8(27-19(34)9(28)4-23)3-7(25)16(14(17)32)38-21-13(31)12(30)11(29)10(5-24)37-21;1-5(2,3)4/h7-18,20-21,26,28-33,35H,3-6,23-25H2,1-2H3,(H,27,34);(H2,1,2,3,4)/t7-,8+,9-,10+,11+,12-,13+,14-,15+,16+,17-,18+,20+,21+,22-;/m0./s1
Chemical Name or Material Isepamicin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.