Learn More
YSI™ ISE Probes for Pro Series Portable Meters
Field probes for ion concentration measurements.
Brand:  YSI™ 605106
Code : 04
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 11999128
  • £348.00 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Requires : Cable/connector assembly
Description
Description
For use with Pro Series Portable Meters.
Specifications
Specifications
For use in fresh water applications
Nitrate ISE
YSI Pro Series Portable Meters