missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISCU Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14812-1-AP-20UL
This item is not returnable.
View return policy
Description
Two ISCU isoforms, ISCU1 (cytosolic, 15 kDa) and ISCU2 (mitochondrial, 18 kDa), were produced by alternative splicing. ISCU2 is considered the canonical isoform 1. Proteintech's ISCU antibody 14812-1-AP is raised against the full-length ISCU2 and is
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.Specifications
| ISCU | |
| Polyclonal | |
| Unconjugated | |
| ISCU | |
| 2310020H20Rik, HML, hnifU, ISCU, ISU2, NIFU, NifU like protein, NIFUN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23479, 288740, 66383 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D7P6, Q9H1K1 | |
| Iscu | |
| ISCU Fusion Protein Ag6562 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction