missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ISCU Monoclonal Antibody (OTI4F5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526595
This item is not returnable.
View return policy
Description
ISCU Monoclonal Antibody for Western Blot, IHC (P)
Iron-sulfur (Fe-S) clusters are necessary for several mitochondrial enzymes and other subcellular compartment proteins. They contain sulfur and iron, and are created via several steps that include cysteine desulfurases, iron donors, chaperones, and scaffold proteins. This gene encodes the two isomeric forms, ISCU1 and ISCU2, of the Fe-S cluster scaffold protein. Mutations in this gene have been found in patients with myopathy with severe exercise intolerance and myoglobinuria.
Specifications
| ISCU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H1K1 | |
| ISCU | |
| Full length human recombinant protein of ISCU produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F5 | |
| Unconjugated | |
| ISCU | |
| 2310020H20Rik; AA407971; HML; hnifU; iron-sulfur cluster assembly enzyme; iron-sulfur cluster assembly enzyme ISCU, mitochondrial; iron-sulfur cluster scaffold homolog; Iscu; IscU iron-sulfur cluster scaffold homolog; IscU iron-sulfur cluster scaffold homolog (E. coli); ISCU2; ISU2; Nifu; NifU-like N-terminal domain containing; nifU-like N-terminal domain-containing protein; NifU-like protein; NIFUN; RGD1309562 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23479 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction