missing translation for 'onlineSavingsMsg'
Learn More

Isaxonine, TRC

Product Code. 30394920
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394920 1 g
30403508 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394920 Supplier Toronto Research Chemicals Supplier No. TRCI9006701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4214-72-6
Molecular Formula C7 H11 N3
Molecular Weight (g/mol) 137.82
Synonym 2-(Isopropylamino)pyrimidine, N-(Isopropylamino)-2-pyrimidine, N-Isopropyl-2-pyrimidinamine, N-(1-Methylethyl)-2-pyrimidinamine, N-Isopropylpyrimidin-2-amine
IUPAC Name N-isopropylpyrimidin-2-amine
SMILES CC(C)Nc1ncccn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H11N3/c1-6(2)10-7-8-4-3-5-9-7/h3-6H,1-2H3,(H,8,9,10)
Formula Weight 137.0953
Chemical Name or Material Isaxonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.