missing translation for 'onlineSavingsMsg'
Learn More

Isavuconazole-d4, TRC

Product Code. 30392271
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392271 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392271 Supplier Toronto Research Chemicals Supplier No. TRCI77783210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isavuconazole-d4
CAS 1346598-58-0
Purity Grade Notes HPLC
Molecular Formula C22H13D4F2N5OS
Quantity 10 mg
Synonym 4-[2-[(1R,2R)-2-(2,5-Difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]-4-thiazolyl]benzonitrile-d4, 2R,3R)-3-[4-(4-Cyanophenyl)thiazol-2-yl]-2-(2,5- difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol-d4, BAL 4815-d4, RO 0094815-d4
InChI Formula InChI=1S/C22H17F2N5OS/c1-14(21-28-20(10-31-21)16-4-2-15(9-25)3-5-16)22(30,11-29-13-26-12-27-29)18-8-17(23)6-7-19(18)24/h2-8,10,12-14,30H,11H2,1H3/t14-,22+/m0/s1/i2D,3D,4D,5D
SMILES FC1=CC([C@](CN2C=NC=N2)(O)[C@@H](C)C3=NC(C4=C([2H])C([2H])=C(C#N)C([2H])=C4[2H])=CS3)=C(F)C=C1
Molecular Weight (g/mol) 441.49
Formula Weight 441.1373
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.