missing translation for 'onlineSavingsMsg'
Learn More

Isatoic anhydride, 97%

Product Code. 11402038
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402038 100 g 100g
11412038 500 g 500g
11422038 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402038 Supplier Thermo Scientific Alfa Aesar Supplier No. A14603.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-48-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00006700
InChI Key TXJUTRJFNRYTHH-UHFFFAOYSA-N
Synonym isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
PubChem CID 8359
SMILES O=C1NC2=CC=CC=C2C(=O)O1

Specifications

Melting Point ∼237°C (decomposition)
Density 1.52
Flash Point 308°C (586°F)
Quantity 100 g
Beilstein 136786
Sensitivity Moisture sensitive
IUPAC Name 2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Formula Weight 163.13
Percent Purity 97%
Chemical Name or Material Isatoic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.