missing translation for 'onlineSavingsMsg'
Learn More

Isatin, MedChemExpress

Product Code. 16429416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
16429416 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 16429416

Brand: Medchem Express HYY0265

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Isatin (Indoline-2,3-dione) is a potent inhibitor of monoamine oxidase (MAO) with an IC50 of 3 μM. Also binds to central benzodiazepine receptors (IC50 against clonazepam, 123 μM). Also acts as an antagonist of both atrial natriuretic peptide stimulated and nitric oxide-stimulated guanylate cyclase activity. Shows effect on the serotonergic system.

Isatin (Indoline-2,3-dione) is a potent inhibitor of monoamine oxidase (MAO) with an IC50 of 3 μM. Also binds to central benzodiazepine receptors (IC50 against clonazepam, 123 μM). Also acts as an antagonist of both atrial natriuretic peptide stimulated and nitric oxide-stimulated guanylate cyclase activity. Shows effect on the serotonergic system.

Specifications

Chemical Name or Material Isatin
CAS 91-56-5
Color Orange to Red
Purity Grade Notes Research
Molecular Formula C8H5NO2
Quantity 100 mg
Synonym Indoline-2,3-dione
Solubility Information DMSO : 110 mg/mL (747.64 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1NC2=C(C=CC=C2)C1=O
Molecular Weight (g/mol) 147.13
Formula Weight 147.13
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.