missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRX6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569494
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 85%; Human: 100%; Mouse: 86%; Rabbit: 92%; Rat: 92%.
IRX6 gene ontology annotations related to this gene include DNA-binding transcription factor activity, RNA polymerase II-specific; sequence-specific DNA binding.
Specifications
| IRX6 | |
| Polyclonal | |
| Unconjugated | |
| IRX6 | |
| homeodomain protein IRXB3; iroquois homeobox 6; iroquois homeobox protein 6; iroquois homeobox protein 7; Iroquois related homeobox 6; Iroquois related homeobox 6 (Drosophila); iroquois-class homeodomain protein IRX-6; IRX-3; IRX6; IRX7; Irxb3; RGD1564830 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 307715 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| IRX6 | |
| synthetic peptide directed towards the middle region of human IRX6. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction