missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRX6 Monoclonal Antibody (1A5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524487
Additional Details : Weight : 0.01000kg
Description
Peptide Sequence: PRLTMHYPCL EKPRIWSLAH TATASAVEGA PPARPRPRSP ECRMIPGQPP ASARRLSVPR DSACDESSCI PKAFGNPKFA LQGLPLNCAP CPRRSEPVVQ CQYPSGAEAG
IRX6 gene ontology annotations related to this gene include DNA-binding transcription factor activity, RNA polymerase II-specific; sequence-specific DNA binding.Specifications
IRX6 | |
Monoclonal | |
Unconjugated | |
IRX6 | |
homeodomain protein IRXB3; iroquois homeobox 6; iroquois homeobox protein 6; iroquois homeobox protein 7; Iroquois related homeobox 6; Iroquois related homeobox 6 (Drosophila); iroquois-class homeodomain protein IRX-6; IRX-3; IRX6; IRX7; Irxb3; RGD1564830 | |
Mouse | |
Protein A | |
RUO | |
79190 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1A5 | |
PBS with no preservative; pH 7.4 | |
P78412 | |
IRX6 | |
IRX6 (NP_077311, 337 a.a. ∽ 446 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |