missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRX3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5111549

Product Code. 17179463

  • £554.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: FARPAEPEGG TDRCSALEVE KKLLKTAFQP VPRRPQNHLD AALVLSALSS Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 86%.

IRX3 is a member of the Iroquois homeobox gene family (seeIRX1; MIM 606197) and plays a role in an early step of neural development. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRX3
Polyclonal
Unconjugated
IRX3
AI894186; homeodomain protein IRXB1; iroquois homeobox 3; iroquois homeobox protein 3; Iroquois related homeobox 3; iroquois-class homeodomain protein IRX-1; iroquois-class homeodomain protein IRX-3; IRX-1; IRX3; Irxb1
Rabbit
Antigen affinity chromatography
RUO
79191
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P78415
IRX3
Recombinant Protein corresonding to Human IRX3. Recombinant protein control fragment (Product #a href=.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.