missing translation for 'onlineSavingsMsg'
Learn More

IRX3 Antibody (2H3), Novus Biologicals™

Product Code. 18404079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18404079 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18404079 Supplier Novus Biologicals Supplier No. H00079191M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRX3 Monoclonal antibody specifically detects IRX3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX3
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2H3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_077312
Gene Alias Homeodomain protein IRXB1, iroquois homeobox 3, Iroquois homeobox protein 3, iroquois-class homeodomain protein IRX-1, iroquois-class homeodomain protein IRX-3, IRX-1, IRXB1FLJ99187
Host Species Mouse
Immunogen IRX3 (NP_077312, 182 a.a. ∽ 285 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RRRLKKENKMTWAPRSRTDEEGNAYGSEREEEDEEEDEEDCKRELELEEEELGGEEEDTGGEGLADDDEDEEIDLENLDGAATEPELSLAGAARRDGDLGLGPI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79191
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.