missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585974
This item is not returnable.
View return policy
Description
IRX2 Polyclonal Antibody for Western Blot, ICC/IF
IRX2 is a member of the Iroquois homeobox gene family. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
Specifications
| IRX2 | |
| Polyclonal | |
| Unconjugated | |
| IRX2 | |
| homeodomain protein IRXA2; iroquois homeobox 2; iroquois homeobox protein 2; Iroquois related homeobox 2 (Drosophila); iroquois-class homeobox protein Irx6; Iroquois-class homeodomain protein IRX-2; Irx2; Irx6; IRXA2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 153572, 16372 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P81066, Q9BZI1 | |
| IRX2 | |
| Synthetic peptide corresponding to Human IRX2 aa 422-471. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction