missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRS4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117329
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
IRS4 encodes the insulin receptor substrate 4, a cytoplasmic protein that contains many potential tyrosine and serine/threonine phosphorylation sites. Tyrosine-phosphorylated IRS4 protein has been shown to associate with cytoplasmic signalling molecules that contain SH2 domains. The IRS4 protein is phosphorylated by the insulin receptor tyrosine kinase upon receptor stimulation.
Specifications
| IRS4 | |
| Polyclonal | |
| Unconjugated | |
| IRS4 | |
| 160 kDa phosphotyrosine protein; Insulin receptor substrate 4; Irs4; IRS-4; Phosphoprotein of 160 kDa; pp160; py160; RP23-415G1.3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8471 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O14654 | |
| IRS4 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human IRS4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction