missing translation for 'onlineSavingsMsg'
Learn More

IRS4 Antibody (5B9), Novus Biologicals™

Product Code. 18378729 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378729 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Produktkod. 18378729 Leverantör Novus Biologicals Leverantörsnummer H00008471M01

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

IRS4 Monoclonal antibody specifically detects IRS4 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen IRS4
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 5B9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003595
Gene Alias insulin receptor substrate 4,160 kDa phosphotyrosine protein, IRS-4pp160, Phosphoprotein of 160 kDa, PY160
Host Species Mouse
Immunogen IRS4 (NP_003595, 850 a.a. ~ 950 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DPKDAASKPSGEGSFSKPGDGGSPSKPSDHEPPKNKAKRPNRLSFITKGYKIKPKPQKPTHEQREADSSSDYVNMDFTKRESNTPAPSTQGLPDSWGIIAE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8471
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.