missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-68244H
This item is not returnable.
View return policy
Description
IRS2 Polyclonal antibody specifically detects IRS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| IRS2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| insulin receptor substrate 2, IRS-2 | |
| The immunogen recognized by this antibody maps to a region between residue 1288 and 1338 of human insulin receptor substrate 2 using the numbering given in entry NP_003740.2 (GeneID 8660). | |
| 0.1 mL | |
| Cell Cycle and Replication, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction | |
| 8660 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction