missing translation for 'onlineSavingsMsg'
Learn More

IRS1 Rabbit anti-Human, Clone: 3Q8A2, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £386.00

Specifications

Antigen IRS1
Clone 3Q8A2
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18310515
View Documents
Novus Biologicals
NBP3-15648-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18344183
View Documents
Bio-Techne
NBP3-15648-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IRS1 Monoclonal antibody specifically detects IRS1 in Human samples. It is validated for Western Blot
Specifications

Specifications

IRS1
Western Blot
Unconjugated
Rabbit
Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Neuroscience, Signal Transduction
PBS, 0.05% BSA, 50% glycerol, pH7.3
3667
IgG
Affinity purified
3Q8A2
Monoclonal
Purified
RUO
Human
HIRS-1, insulin receptor substrate 1, IRS-1
A synthetic peptide corresponding to a sequence within amino acids 1100-1200 of human IRS1 (P35568).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.