missing translation for 'onlineSavingsMsg'
Learn More

IRS1 Antibody (OTI3G10), DyLight 594, Novus Biologicals™

Product Code. 18407474 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407474 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407474 Supplier Novus Biologicals Supplier No. NBP271049DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRS1 Monoclonal antibody specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRS1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3G10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias HIRS-1, insulin receptor substrate 1, IRS-1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-280 of human IRS1 (NP_005535) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3667
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.