missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | IRP2 |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18372044
|
Novus Biologicals
NBP3-03393-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18308476
|
Novus Biologicals
NBP3-03393-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRP2 Polyclonal specifically detects IRP2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| IRP2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| ACO3, EC 4.2.1.3, FLJ23381, IRE-BP 2, Iron regulatory protein 2, iron-responsive element binding protein 2, iron-responsive element-binding protein 2, IRP2IRP2AD | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human IRP2 (NP_004127.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| HIF Target Genes, Hypoxia, Lipid and Metabolism | |
| PBS with 50% glycerol, pH7.3. | |
| 3658 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title