missing translation for 'onlineSavingsMsg'
Learn More

Iroquois-class homeodomain protein IRX-5 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-21441JF525

Product Code. 30175840

  • £343.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Iroquois-class homeodomain protein IRX-5 Polyclonal antibody specifically detects Iroquois-class homeodomain protein IRX-5 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Iroquois-class homeodomain protein IRX-5
Polyclonal
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
10265
Store at 4°C in the dark.
IgG
Immunocytochemistry/Immunofluorescence
Janelia Fluor 525
Homeodomain protein IRX-2A, Homeodomain protein IRXB2, iroquois homeobox 5, Iroquois homeobox protein 5, iroquois-class homeodomain protein IRX-5, IRX-2a, IRX2A, IRXB2
Produced in rabbits immunized with E. coli-derived Human Iroquois-class homeodomain protein IRX-5 fragment
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.