missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iroquois-class homeodomain protein IRX-5 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21441G
This item is not returnable.
View return policy
Description
Iroquois-class homeodomain protein IRX-5 Polyclonal antibody specifically detects Iroquois-class homeodomain protein IRX-5 in Human samples. It is validated for Immunofluorescence
Specifications
| Iroquois-class homeodomain protein IRX-5 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| Homeodomain protein IRX-2A, Homeodomain protein IRXB2, iroquois homeobox 5, Iroquois homeobox protein 5, iroquois-class homeodomain protein IRX-5, IRX-2a, IRX2A, IRXB2 | |
| Produced in rabbits immunized with E. coli-derived Human Iroquois-class homeodomain protein IRX-5 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10265 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction