missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iroquois-class homeodomain protein IRX-5 Antibody (6A9), Novus Biologicals™
Shop All Bio Techne ProductsDescription
Iroquois-class homeodomain protein IRX-5 Monoclonal antibody specifically detects Iroquois-class homeodomain protein IRX-5 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Iroquois-class homeodomain protein IRX-5 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 6A9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005844 |
| Gene Alias | Homeodomain protein IRX-2A, Homeodomain protein IRXB2, iroquois homeobox 5, Iroquois homeobox protein 5, iroquois-class homeodomain protein IRX-5, IRX-2a, IRX2A, IRXB2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?