missing translation for 'onlineSavingsMsg'
Learn More

Iron wire, 0.1mm (0.004in) dia, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number 11341488
Click to view available options
Quantity:
10 m
50 m
This item is not returnable. View return policy
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]
This item is not returnable. View return policy
7439-89-6
Fe
55.85
MFCD00010999
XEEYBQQBJWHFJM-UHFFFAOYSA-N
ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
23925
CHEBI:82664
iron
[Fe]

Form: approx. 0.06g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray wire
Density 7.87g/cm3 at 20°C
Assay Percent Range 99.998% (Metals basis)
Quantity 10 m
Form Wire
Chemical Name or Material Iron

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'