missing translation for 'onlineSavingsMsg'
Learn More

Iron Trisglycinate, TRC

Product Code. 30393712
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393712 1 g
30395242 500 mg
30391216 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393712 Supplier Toronto Research Chemicals Supplier No. TRCI7754751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34369-82-9
Molecular Formula C6H12FeN3O6
Molecular Weight (g/mol) 278.02
Synonym Ferric Glycinate, Iron Glycinate, Tris(glycinato)iron(III)
IUPAC Name 2-aminoacetate;iron(3+)
SMILES [Fe+3].NCC(=O)[O-].NCC(=O)[O-].NCC(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/3C2H5NO2.Fe/c3*3-1-2(4)5;/h3*1,3H2,(H,4,5);/q;;;+3/p-3
Formula Weight 278.0076
Chemical Name or Material Iron Trisglycinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.