missing translation for 'onlineSavingsMsg'
Learn More

Iron sulfide, 99.9% (metals basis)

Product Code. 11327577
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 11327577

Brand: Thermo Scientific Alfa Aesar 012842.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12068-85-8 | FeS2 | 119.97 g/mol

Iron sulfide is used in photovoltaics and lithium ion battery cathodes. It is also studied as a potential material for the sequestration of environmental contaminants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12068-85-8
Molecular Formula FeS2
Molecular Weight (g/mol) 119.97
MDL Number MFCD00064690
InChI Key NFMAZVUSKIJEIH-UHFFFAOYSA-N
Synonym iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
PubChem CID 14788
SMILES S=[Fe]=S

Specifications

Melting Point 1171°C
Color Gray
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name disulfanylideneiron
Formula Weight 119.98
Odor Odorless
Appearance Gray powder
Density 4.87 g/cm3
Chemical Name or Material Iron sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.