missing translation for 'onlineSavingsMsg'
Learn More

Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 196051000

Code : 29

Additional Details : CAS Number : 7732-18-5 Weight : 0.15000kg Transport : UN number : 3264 Chem class : 8 Pack group : III


 View more versions of this product

Product Code. 10103740

  • £25.60 / 100mL
Estimated Shipment: 25-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Iron standard solution
1 mg/ml Fe in 2% HNO3
Plastic bottle
Colorless
MFCD00010999
[Fe]
55.85
CHEBI:82664
AAS
Liquid
997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C)
7439-89-6,7697-37-2,7732-18-5
100 mL
Fe
XEEYBQQBJWHFJM-UHFFFAOYSA-N
iron
23925
55.85
7.86
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iron standard solution

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only