missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

Product Code. 10533142 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10533142 100 mL 100mL
10090190 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533142 Supplier Fisher Chemical Supplier No. J/8030/05
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10421-48-4
Molecular Formula FeN3O9
Molecular Weight (g/mol) 241.857
MDL Number 11003
InChI Key VCJMYUPGQJHHFU-UHFFFAOYSA-N
Synonym ferric nitrate, iron trinitrate, nitric acid, iron 3+ salt, unii-n8h8402xob, iron nitrate fe no3 3, iron iii nitrate, anhydrous, hsdb 451, iron 3+ trinitrate, nitric acid, iron 3+ salt 3:1
PubChem CID 25251
IUPAC Name iron(3+);trinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3]

Specifications

Color Colorless
pH 1
Quantity 100 mL
Packaging HDPE plastic bottle
Form Liquid
compliance-icons
Product Identifier
  • Iron solution 1000 ppm
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Nitric acid, Ferricnitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.