missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Standard for Ion Chromatography (IC) 1 mg/mL (1000 ppm) in 0.005% Nitric Acid (HNO3), Reagecon
Shop All Reagecon Products

Description
- Ion Chromatography Reference Standard, Ion Chromatography Reference Material
- ISO/IEC 17025 for the testing of ICP/ICP-MS Standards using ICP-MS (INAB Ref: 264T)
- ISO/IEC 17025 for calibration of laboratory balances (INAB Ref: 265C)
- ISO/IEC 17025 for temperature calibration in the range of -196 - +1200°C (INAB Ref: 265C)
- ISO/IEC 17025 for density measurement (INAB Ref: 264T) using Oscillating U-Tube and Bingham Pycnometry
- ISO/IEC 17025 for certain complexometric titrations (INAB Ref: 264T)
- GHS/OSHA compliant, REACH etc.
- This product is produced in a highly controlled cleanroom (ISO 7) environment, using ultra-pure water (specially treated for the production of ion chromatography standards) and triple leached pre-cleaned bottles.
- The titrimetry and spectroscopy are controlled and calibrated using separate Certified Reference Materials (CRM’s), so the product comes with two layers of traceability including NIST Standards where available.
- All of the uncertainties of measurement for the product are calculated according to Eurachem/CITAC guidelines and reported as expanded uncertainties at the 95% confidence level.
Specifications
Specifications
| Description | Matrix: 0.005% HNO₃ |
| Packaging | Amber Glass Triple-leached |
| Analyte or Component Names | Iron |
| Physical Form | Liquid |
| Concentration | 1 mg/mL (1000 ppm) |
| Technique | Ion Chromatography, Cation Measurement |
| Element | Fe |
| Quantity | 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?