missing translation for 'onlineSavingsMsg'
Learn More

Iron silicide, 99.9% (metals basis)

Product Code. 11341549
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341549 100 g 100g
11331549 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341549 Supplier Thermo Scientific Alfa Aesar Supplier No. 014019.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12022-95-6 | FeSi | 83.93 g/mol

Iron silicide is used in deoxidation and alloying of steel melts, alloying of cast iron charges and melts (Iron foundries), silicothermic reduction of precious metals (chromium, molybdenum and vanadium) and others (Nb, W), Production of some inoculants and nodulisers by alloying (melting). High purity iron silicide is used to produce high permeability steel for electric transformers, production of FeSiMn, FeSiN and SiMn- alloys. It is also used as metal filler.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12022-95-6
Molecular Formula FeSi
Molecular Weight (g/mol) 83.93
MDL Number MFCD00016086
InChI Key YSWDZWXHBYTMBR-UHFFFAOYSA-N
Synonym silyliron, silylene iron, ferric silicon, silanidylidyneironylium
PubChem CID 92027036
SMILES [Si-]#[Fe+]

Specifications

Melting Point 1410°C
Color Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name silanidylidyneiron(1+)
Formula Weight 83.93
Odor Odorless
Appearance Gray powder
Chemical Name or Material Iron silicide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.