missing translation for 'onlineSavingsMsg'
Learn More

Iron Saccharate, TRC

Product Code. 30391217
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391217 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391217 Supplier Toronto Research Chemicals Supplier No. TRCI77545050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron Saccharate
CAS 8047-67-4
Molecular Formula [Na2Fe5O8(OH) • 3H2O]n •(C12H22O11)m
Quantity 50 g
Synonym Colliron IV, Feojectin, Ferric Hydroxide Sucrose Complex, Ferrivenin, Ferrum Hausmann, Ferrum Vitis, Ferum Hausmann, Fesin, Hippiron, Iron Oxide Saccharated, Iron Sucrose, Iron Sucrose Complex, Iron Sugar, Iron(III) Hydroxide Sucrose Complex, Iviron, Neoferrum, Proferrin, Saccharated Ferric Oxide, Saccharated Iron, Saccharated Iron Oxide, Sucrofer, Venofer, Venoferrum, XI 921
InChI Formula InChI=1S/3C6H10O8.2Fe/c3*7-1(3(9)5(11)12)2(8)4(10)6(13)14;;/h3*1-4,7-10H,(H,11,12)(H,13,14);;/q;;;2*+3/p-6/t3*1-,2-,3-,4+;;/m000../s1
SMILES [C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[Fe+3].[Fe+3]
Molecular Weight (g/mol) 736.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.